Learn More
Medchemexpress LLC AFM32a hydrochloride | 2988594-85-8 | 99.4% | 517.00 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY136557A
Encompass_Preferred
This benzimidazole-based derivative is a potent and selective protein arginine deiminase 2 inhibitor. It exhibits superior selectivity for PAD2 over PAD4 (95-fold) and PAD3 (79-fold).
  • Potent and selective protein arginine deiminase 2 inhibitor.
  • Superior selectivity for PAD2 over PAD4 (95-fold) and PAD3 (79-fold).
Catalog No. 50-003-47894