Learn More
Cayman Chemical AFMK Neurochemicals
SDP
Supplier:  Cayman Chemical 100052545 MG
Encompass_Preferred
An antioxidant metabolite of melatonin; first identified in rat brain; has antioxidant and free radical scavenging activities; may be measured in plasma as an index of melatonin synthesis and metabolism
Catalog No. 50-288-7963