Learn More
Selleck Chemical LLC AFN-1252-E1126-5MG
SDP
Supplier:  Selleck Chemical LLC E11265MG
Encompass_Preferred
AFN-1252 (Debio 1452) is a potent inhibitor of enoyl-acyl carrier protein reductase (FabI) inhibits all clinical isolates of Staphylococcus aureus and Staphylococcus epidermidis at concentrations of 0 12 g/ml
Catalog No. 50-313-5662