Learn More
Genscript Corporation AFP RC6B mAb Mouse
Supplier:  Genscript Corporation V063021
Encompass
Alpha-Fetoprotein (AFP) is a glycoprotein normally produced during fetal development by the hepatocytes Its level is increased in pregnant women AFP is an important marker for the diagnosis of hepatocellular carcinoma
Catalog No. 50-000-57052