Learn More
TARGETMOL CHEMICALS INC AFURESERTIB HYDROCHLORID 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T788525MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Afuresertib hydrochloride is an inhibitor of Akt kinase (Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively)purity: 100%
Catalog No. 50-257-9112