Learn More
Medchemexpress LLC Afuresertib hydrochloride | 1047645-82-8 | 100.0% | 463.78 | C18H18Cl3FN4OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15727A10MG
Encompass_Preferred
Afuresertib hydrochloride is the hydrochloride salt of afuresertib, a selective, ATP-competitive pan-Akt kinase inhibitor used in biochemical and cellular research to probe PI3K/Akt signaling. Supplied as a solid (white to yellow), the product is intended for research use only and is provided in small, accurately measured quantities for laboratory experiments.
  • Research-grade compound with high listed purity (99.95%).
  • Solid form, white to yellow, suitable for storage and handling in typical laboratory conditions.
  • Used to inhibit Akt kinase activity in biochemical and cell-based assays.
  • Available in small mass vials (common size: 10 mg) and solution aliquots for convenience.
  • Provided with supporting product documentation including a datasheet with physicochemical properties.
Catalog No. 50-002-87715