Learn More
Selleck Chemical LLC Afuresertib hydrochloride E4992-5MG
SDP
Supplier:  Selleck Chemical LLC E49925MG
Encompass_Preferred
Afuresertib hydrochloride (GSK 2110183 hydrochloride) is an orally bioavailable selective ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0 08/2/2 6 nM for Akt1/Akt2/Akt3 respectively
Catalog No. 50-000-41176