Learn More
Cayman Chemical AfuresertIbhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1798810 MG
Encompass_Preferred
A pan-Akt inhibitor (IC50s = 0.08, 2, and 2.6 nM for Akt1, -2, and -3, respectively); selective for Akt over a panel of 13 kinases (IC50s = >100 nM) but does inhibit PKA, PKG1α, and PKG1β (IC50s = 1.3, 0.9, and 4 nM, respectively); inhibits tumor growth in an SKOV3 mouse xenograft model at 10, 30, and 100 mg/kg
Catalog No. 50-296-5429