Learn More
Selleck Chemical LLC AG-1024 S1234-5mg
SDP
Supplier:  Selleck Chemical LLC S12345MG
Encompass_Preferred
AG-1024 (Tyrphostin AGS 200) inhibits IGF-1R autophosphorylation with IC50 of 7 M is less potent to IR with IC50 of 57 M and specifically distinguishes between InsR and IGF-1R (as compared to other tyrphostins)
Catalog No. 50-310-8107