Learn More
Selleck Chemical LLC Ivosidenib (AG-120) 5mg 1448347-49-6
SDP
Supplier:  Selleck Chemical LLC S82065MG
Encompass_Preferred
Ivosidenib (AG-120) is an orally available inhibitor of isocitrate dehydrogenase type 1 (IDH1), with potential antineoplastic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5803