Learn More
TARGETMOL CHEMICALS INC AG 1295 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1413725MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. AG 1295, a selective inhibitor of platelet-derived growth factor receptor (PDGFR) tyrosine-kinase, effectively halts PDGFR autophosphorylation without impacting the autophosphorylation of the EGF receptor[1][2][3][4]. Purity 99.64%
Catalog No. 50-288-0478