Learn More
Selleck Chemical LLC AG-13958 S2211-2mg
SDP
Supplier:  Selleck Chemical LLC S22112MG
Encompass_Preferred
AG-13958 (AG-013958) a VEGFR tyrosine kinase inhibitor is in clinical development with ST administration for treatment of choroidal neovascularization associated with age related macular degeneration (AMD)
Catalog No. 50-311-2703