Learn More
Medchemexpress LLC HY-15492 5mg Medchemexpress, AG-13958 CAS:319460-94-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY154925MG
Encompass_Preferred
Medchemexpress, HY-15492 5mg AG-13958 CAS:319460-94-1 AG-13958 (AG-013958), a potent VEGFR tyrosine kinase inhibitor, is used for treatment of choroidal neovascularization associated with age-related macular degeneration (AMD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9943