Learn More
Selleck Chemical LLC AG-1478 S2728-25mg
SDP
Supplier:  Selleck Chemical LLC S272825MG
Encompass_Preferred
AG-1478 is a selective EGFR inhibitor with IC50 of 3 nM in cell-free assays almost no activity on HER2-Neu PDGFR Trk Bcr-Abl and InsR AG-1478 (Tyrphostin AG-1478) inhibits encephalomyocarditis virus (EMCV) and hepatitis c virus (HCV) by targeting phosphatidylinositol 4-kinase III (PI4KA)
Catalog No. 50-311-1817