Learn More
Selleck Chemical LLC AG-1478 (Tyrphostin AG-1478) 10mM/1mL 153436-53-4 NSC 693255
SDP
Supplier:  Selleck Chemical LLC S272810MM/1ML
Encompass_Preferred
AG-1478 (Tyrphostin AG-1478, NSC 693255) is a selective EGFR inhibitor with IC50 of 3 nM in cell-free assays, almost no activity on HER2-Neu, PDGFR, Trk, Bcr-Abl and InsR. AG-1478 (Tyrphostin AG-1478) inhibits encephalomyocarditis virus (EMCV) and hepatitis c virus (HCV) by targeting phosphatidylinositol 4-kinase III? (PI4KA). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6208