Learn More
Medchemexpress LLC AG-270 | 2201056-66-6 | 99.97% | 489.57 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13863050MG
Encompass_Preferred
AG-270 is a first-in-class, allosteric, noncompetitive, reversible, and orally active MAT2A inhibitor with an IC50 of 14 nM. It potently reduces intracellular S-adenosylmethionine (SAM) levels and demonstrates MTAP-null-selective antiproliferative activity. In vivo, AG-270 exhibits excellent metabolic stability and leads to dose-dependent reductions in tumor SAM levels and tumor growth in xenografts. Its combination with taxanes and gemcitabine has shown additive-to-synergistic antitumor activity.
  • Potent and selective MAT2A inhibitor
  • Reduces intracellular SAM levels
  • Shows MTAP-null-selective antiproliferative activity
  • Excellent metabolic stability across multiple species
  • Exhibits dose-dependent reduction in tumor SAM levels and tumor growth
  • Additive-to-synergistic antitumor activity in combination with other therapies
Catalog No. 50-003-50779