Learn More
Selleck Chemical LLC AG-270 S9872-25MG
SDP
Supplier:  Selleck Chemical LLC S987225MG
Encompass_Preferred
AG-270 is a potent and non-competitive methionine adenosyltransferase 2A (MAT2A) inhibitor which lessens the intracellular SAM levels and MTAP-null selective antiproliferative activity
Catalog No. 50-000-24672