Learn More
Enzo Life Sciences AG-494 (10mg). CAS: 133550-35-3
SDP
Supplier:  Enzo Life Sciences BMLEI2280010
Encompass_Preferred
AG-494 is a member of the tyrphostin family of tyrosine kinase inhibitors and is a potent inhibitor of EGF receptor autophosphorylation (IC50=1.2 µM) and EGF-dependent cell growth (IC50=6 µM). It selectively inhibits HER1 (EGF receptor) vs. HER1-2 receptor autophosphorylation (HER1: IC50=1.1 µM; HER1-2: IC50=45 µM). HER1-2 is a chimeric receptor consisting of the external HER1 domain fused to an internal HER2 domain. Blocks CDK2 activation and causescells to arrest at late G1 and during S phase. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0620