Learn More
Selleck Chemical LLC AG-636 S9776-25MG
SDP
Supplier:  Selleck Chemical LLC S977625MG
Encompass_Preferred
AG-636 is an orally available inhibitor of dihydroorotate dehydrogenase (DHODH) with potential antineoplastic activity
Catalog No. 50-000-23979