Learn More
Medchemexpress LLC HY-15844 5mg Medchemexpress, AG-825 CAS:149092-50-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY158445MG
Encompass_Preferred
Medchemexpress, HY-15844 5mg AG-825 CAS:149092-50-2 AG-825 (Tyrphostin AG-825) is a selective and ATP-competitive ErbB2 inhibitor which suppresses tyrosine phosphorylation, with an IC50 of 0.35 μM. AG-825 displays anti-cancer activity. AG825 significantly accelerates apoptosis of human neutrophils[4]. AG-825 is a potential agent for overcoming Mn-induced neurotoxicity or AD development[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1804