Learn More
TARGETMOL CHEMICALS INC AG1024 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T269325MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AG1024 (Tyrphostin) suppresses IGF-1R autophosphorylation(IC50=7 uM) and is less potent for IR(IC50=57 uM). purity: 99%
Catalog No. 50-258-0663