Learn More
TARGETMOL CHEMICALS INC AG14361 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T63395MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AG14361 is an effective inhibitor of PARP1 (Ki<5 nM). purity: 99%
Catalog No. 50-257-9425