Learn More
Cayman Chemical AG-1478, 153436-53-4, 1 mg
SDP
Supplier:  Cayman Chemical 100102441
Encompass_Preferred
Molecular Formula C16H14ClN3O2, Purity ≥98%, Formula Weight 315.8, Synonyms: N-(3-chlorophenyl)-6,7-dimethoxy-4-quinazolinamine, NSC 693255, Tyrphostin AG-1478; An inhibitor of EGFR kinase with an IC50 value of 3 nM.
Catalog No. NC0053836
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.