Learn More
Cayman Chemical AG-490, 133550-30-8, 10 mg
SDP
Supplier:  Cayman Chemical 1001031110
Encompass_Preferred
Molecular Formula C17H14N2O3, Purity ≥98%, Formula Weight 294.3, Synonyms: 2-cyano-3-(3,4-dihydroxyphenyl)-N-(phenylmethyl)-2-propenamide, Tyrphostin AG-490; An selective inhibitor of JAK2 activity that blocks leukemic cell growth in vitro and in vivo by inducing programmed cell death; blocks growth of all pre-B acute leukemia cells at a concentration of 5 M.
Catalog No. NC9915522
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.