Learn More
Medchemexpress LLC Ag490 (Tyrphostin AG490) | 133550-30-8 | 99.9% | 200 MG
SDP
Supplier:  Medchemexpress LLC HY12000200MG
Encompass_Preferred
AG490 (Tyrphostin AG490) is a small-molecule tyrosine kinase inhibitor that targets JAK2, JAK3, and EGFR, blocking downstream STAT activation. It is supplied for research use and is available as a solid and as DMSO solution for in vitro biochemical and cell-based assays.
  • Inhibits JAK2/3 and EGFR-mediated STAT signaling.
  • Suitable for in vitro kinase inhibition and cell signaling studies.
  • High analytical purity reported at 99.86%.
  • Available as a solid and as a DMSO solution for assay preparation.
  • CAS number 133550-30-8 supports unambiguous identification.
Catalog No. 50-002-33275