Learn More
Cayman Chemical AG879 Inhibitors
SDP
Supplier:  Cayman Chemical 1079310 MG
Encompass_Preferred
A tyrphostin compound that inhibits nerve growth factor (NGF)-dependent TrkA tyrosine phosphorylation in PC-12 cells (IC50 = ~40μM), HER2-ErbB2 kinase in several breast and ovarian cancer cell lines (IC50 = ~0.5 μM), and the VEGF receptor FLK1 (IC50 = ~1 μM)
Catalog No. 50-289-0318