Learn More
LabScientific Agar, Bacteriological | 5kg, Purest form of general purpose agar available. Drosophila.
Supplier:  LabScientific A466
Encompass
Agar, bacteriological grade, 5kg. Supports the growth of a wide variety of both fastidious and non-fastidious organisms. Purest general purpose agar available.
Catalog No. NC9059435