Learn More
Alkali Scientific Agar, Bacteriological Grade, 5 Kg
SDP
Supplier:  Alkali Scientific AGB021CP
Encompass_Preferred
Agar, Bacteriological Grade, 5 Kg is a high-purity agar powder designed for microbiological media preparation. This larger 5 Kg pack size is perfect for laboratories that require larger quantities of agar for routine microbiological analysis and media preparation. Bacteriological grade agar provides a solid, stable surface that supports the growth of microorganisms, helping researchers and laboratory technicians isolate and study cultures under controlled conditions. This powder form is easy to mix and provides reliable gel strength. It is widely used in clinical, research, and industrial applications to cultivate and identify bacteria, fungi, and other microorganisms.
Catalog No. 50-006-60422