Learn More
Bioworld Agar, Low Melt, 1 kg
Supplier:  Bioworld 401000482
Encompass_Preferred
AgarAllows enhanced growth of organisms to optimum levels and offers better solubility of lower temperaturesGel Strength: >1100 g/cm2 Melting Temperature: 85 °CAsh Content: 2.5%Soluble in waterThis product is for laboratory research use only
Catalog No. 50-255-200