Learn More
Medchemexpress LLC Agar, microbiology tested, for cell culture (powder) | 9002-18-0 | MFCD00081288 | 336.33 g/mol | C14H24O9 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW13442350G
Encompass_Preferred
Agar, microbiology tested, for cell culture is a microbiology-grade gelling agent derived from red algae, supplied as a granular powder for preparing solid media and gels in microbial and select cell-culture applications. It forms thermoreversible gels and is suitable for laboratory media preparation, tissue culture support, and analytical assays.
  • Microbiology-tested for suitability in culture media.
  • Forms thermoreversible gels with high gel strength.
  • Powder form for accurate weighing and easy dissolution.
  • Low ash and low insoluble impurities for reproducible results.
  • Available in small research-scale package sizes.
Catalog No. 50-002-82767