Learn More
Sigma Aldrich Fine Chemicals Biosciences Agar suitable for plant ce500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A7921500G
Encompass
Agar contains agarose and agaropectin with agarose being the major gelling heteropolysaccharide. Agarose contains -d-galactose and 3 6-anhydro -l-galactose linked by glycosidic bonds (1-4) called the neoagarobiose. It also has agarobiose with same disaccharide units with(1-3) linkage. The major source of agar is the cell wall of seaweed from Rhodophyceae family.
Catalog No. 50-290-3535