Learn More
Sigma Aldrich Fine Chemicals Biosciences Agarose for molecular biol100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A4718100G
Encompass
Agarose is a polymer extracted from agar or agar-bearing marine algae. This purified linear galactan hydrocolloid comprises alternating co-polymers D-galactose and 3 6-anhydro-L-galactose units connected by (1A†3) and -(1A†4) glycosidic bonds. Agarose is highly biocompatible and possesses variable mechanical and diffusion properties. Agarose can be used as a gelling agent to separate nucleic acids electrophoretically.
Catalog No. 50-290-3754