Learn More
Alkali Scientific Agarose, Low EEO, 100 G
SDP
Supplier:  Alkali Scientific AGA007CP
Encompass_Preferred
Agarose, Low EEO, 100 G is a molecular biology grade agarose that features a low electroendosmosis (EEO) property, which helps to minimize the migration of ions in the gel during electrophoresis. This makes it an ideal choice for applications that require low background interference, such as high-resolution gel electrophoresis. The 100g package is perfect for mid-sized laboratories that require a moderate volume of agarose for regular gel preparations. The low EEO property ensures better band clarity and sharper separation of nucleic acids and proteins, making it ideal for applications where precision is essential, such as in nucleic acid analysis and protein assays.
Catalog No. 50-006-60165