Learn More
Vector Laboratories Succinylated Wheat Germ Agglutinin (WGA), Agarose bound, 2mL
SDP
Supplier:  Vector Laboratories AL1023S2
Encompass
Bead diameter ranges in size from 45-165 microns. Matrix is stable in solutions at pH 3-11 as well as many organic solvents. Immobilized lectins are prepared using affinity purified lectins
Catalog No. NC1695033