Learn More
Medchemexpress LLC HY-N1468 5mg Medchemexpress, Agarotetrol CAS:69809-22-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN14685MG
Encompass_Preferred
Medchemexpress, HY-N1468 5mg Agarotetrol CAS:69809-22-9 Agarotetrol is a chromone derivative isolated from Agarwood. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3838