Learn More
Medchemexpress LLC Agatolimod | 525625-52-9 | 98.8% | 7699.00 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1462441MG
Encompass_Preferred
Agatolimod is a potent TLR9 agonist and immunomodulator, active against human TLR9 with an EC50 of 180 nM. It functions by activating and upregulating both TLR9 and TLR6, mediating downstream signaling pathways. This compound induces Th1-type innate and adaptive immune responses, making it a valuable tool for various research applications including COVID-19, breast cancer, and melanoma.
  • Induces Th1-type innate and adaptive immune responses
  • Activates various immune cells and promotes antigen presentation
  • Upregulates the secretion of multiple cytokines
  • Enhances cytotoxicity and clears intracellular Salmonella
  • Increases radiosensitivity in certain cell lines
  • Enhances anti-spike IgG and neutralizing antibody titers
Catalog No. 50-003-63856