Learn More
Medchemexpress LLC Agatolimod (ODN 2006) | 525625-52-9 | 96.7% | 7699.00 g/mol | C238H314N75O127P23S23 | 5 MG

Supplier: Medchemexpress LLC HY1462445MG
Agatolimod (ODN 2006) is a class B oligodeoxynucleotide TLR9 agonist containing an optimized CpG motif. It is used as a research reagent to stimulate innate immune responses, induce cytokine production, and support preclinical immuno-oncology studies.
- Sequence: 5'-tcgtcgttttgtcgttttgtcgtt-3'.
- Molecular weight: 7,699 g/mol (manufacturer value).
- Purity: 96.73% (product-level listing).
- Solubility: water ≥125 mg/mL.
- Storage: sealed, away from moisture; short-term 4°C; in solvent -80°C for long term.
- Applications: TLR9 activation, cytokine induction, preclinical immuno-oncology research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.