Learn More
Adam Equipment AGB 16a Bench Scale 16lb / 8kg capacity x 0.0005lb / 0.2g readability
Supplier:  Adam Equipment AGB 16A
Encompass_Preferred
Adam Equipment AGB Bench Scales are Ideal for industrial weighing and counting, AGB and AGF bench and floor scales feature sturdy construction and a rugged stainless steel indicator. The scales are available in a choice of platform sizes and a wide selection of capacities. Operation is fast and easy, as results are readily visible on the backlit LCD that boasts 1.5in / 40mm-high digits. An internal rechargeable battery means the AGB/AGF can be used in locations where power is unavailable. Swiftly transmit results to computers or printers using the RS-232 interface. Versatile and easy to operate, AGB and AGF scales are well-suited for use in warehouses, shipping and receiving areas, factories and production facilities. The scales offer capacities from 8kg to 600kg and perform a variety of weighing and counting tasks. Featuring an internal rechargeable battery, the AGB/AGF can be used in locations where power is unavailable. Rechargeable battery included for operation almost anywhere.
Catalog No. NC2611855