Learn More
AdipoGen AGD
SDP
Supplier:  AdipoGen CDXD0169M250
Encompass_Preferred
Chemical. CAS 1456890-74-6. Formula C18H24N2O6. MW 364.39. Synthetic. Coumarin-based turn-off fluorescent probe. Highly selective for ferrous ions Fe2 and can reversibly detect them in aqueous medium. The probe localizes to the cell membrane in living cells, where it can detect changes in Fe2 concentration. Molecular dynamics MD simulations indicate that AGD interacts with the lipid bilayer at the level of the glycerol moieties. Spectral data Excitation=430nm Emission=480nm HEPES, pH 7.4 at RT.
Catalog No. 50-256-1204