Learn More
Selleck Chemical LLC Agerafenib
SDP
Supplier:  Selleck Chemical LLC S80155MG
Encompass_Preferred
Agerafenib (CEP-32496) is a highly potent inhibitor of BRAF(V600E/WT) and c-Raf with Kd of 14 nM/36 nM and 39 nM also potent to Abl-1 c-Kit Ret (c-Ret) PDGFR and VEGFR2 respectively insignificant affinity for MEK-1 MEK-2 ERK-1 and ERK-2 Phase 1/2
Catalog No. 50-000-03888