Learn More
Medchemexpress LLC AGI-24512 | 2201066-53-5 | 99.5% | 400.47 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY112130
Encompass_Preferred
AGI-24512 is a potent methionine adenosyltransferase 2α (MAT2A) inhibitor with an IC50 of 8 nM. It triggers a DNA damage response and can block the proliferation of MTAP-deleted cancer cells in vitro. This compound is used for researching anticancer applications.
  • Potent MAT2A inhibitor with an IC50 of 8 nM.
  • Triggers DNA damage response.
  • Blocks proliferation of MTAP-deleted cancer cells.
  • Used for researching anticancer properties.
Catalog No. 50-004-38816