Learn More
Chemglass Life Sciences Agitator Assembly for 15 L Bioreactors, 15 mm Shaft (CLS-1385-05)
SDP
Supplier:  Chemglass Life Sciences CLS138505
Encompass
Agitator assembly engineered for 15 L bioreactors, providing a contamination-resistant shaft connection and secure mounting to the reactor headplate. Suitable for microbial and cell-culture mixing where stainless-steel construction and reliable sealing are required.
  • Constructed from 316L stainless steel for corrosion resistance.
  • 15 mm shaft outer diameter compatible with standard impellers.
  • 392 mm shaft length through the headplate for deep mixing applications.
  • Mounts to the headplate with four bolts and screws into an M30 headplate thread for secure installation.
Catalog No. 50-273-3309