Learn More
Chemglass Life Sciences Agitator, PTFE, High Viscosity, 8in Diameter, Fits 19mm Shaft Size, Fits 20L Reactors
Supplier: Chemglass Life Sciences CG209610
Agitator, PTFE, High Viscosity, 8in Diameter, Fits 19mm Shaft Size, Fits 20L Reactors. High viscosity agitator, made entirely of PTFE and glass filled PTFE, provides excellent stirring at the bottom of 20L reactors, especially when heavy slurries are being mixed. Agitator attaches to stir shaft by both a compression fitting and a glass filled PTFE safety pin. Heavy duty blades are 7mm thick. Supplied complete with one glass filled PTFE safety pin.