Learn More
Ace Glass, Inc. Agitator, turbine, PTFE, 10mm OD, 64mm diameter
SDP
Supplier:  Ace Glass, Inc. 809008
Encompass_Preferred
Agitator, turbine, PTFE, 10mm, OD, 64mm diameter.All-PTFE turbine type agitator. Unique locking arrangement assures positive grip. Agitator fits 10 mm shafts. Shaft opening is drilled straight thru to allow positioning of agitator at any drilled position on shaft as well as the bottom. For use with bead type glass shafts, 8068-G, 8075 or PTFE shaft, 8071.
Catalog No. NC1932332