Learn More
Gold Biotechnology Inc AGL1 Agrobacterium Chemically Competent Cells 5x50µL
SDP
Supplier:  Gold Biotechnology Inc CC1065X50
Encompass_Preferred
GoldBio’s AGL-1 Agrobacterium chemically competent cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our AGL-1 strain harbors the C58 chromosomal backbone with an insertion mutation in its recA recombination gene. This mutation stabilizes recombinant plasmids. AGL-1 also has rifampicin and carbenicillin resistance genes in the genome useful for selection. A functional T-DNA binary system can be built using our AGL-1 strains as the T-DNA region has been deleted from the Ti plasmid pTiBO542 and instead it has a binary vector containing the missing T-region. The binary system makes possible to transfer genetic material into a host plant S genome. Therefore, our system is often used for
Catalog No. NC2893157