Learn More
Lonza Walkersville AGM Astrocyte Growth Medium SingleQuots Supplements and Growth Factors
Supplier:  Lonza Walkersville CC4123
Encompass
Clonetics AGM Astrocyte Growth Medium has been developed to support the growth of primary human astrocytes. Clonetics Media Products are provided as convenient BulletKits which contain basal medium and necessary growth factors.Basal medium and SingleQuots Kit growth factors are available separately. Clonetics Media Prodcuts are specially designed to support the growth of human and animal primary cells. Media systems have been specifically developed and optimized for various cell systems.Media are supplied in several formats:Basal Medium - contains no growth factors cytokines, or supplementsSingleQuots - growth factors, cytokines, and supplementsBulletKits - basal media and SingleQuots Kit packaged togetherContent:Kit contains growth supplements for a complete growth medium, developed especially for astrocytes.
Catalog No. NC0738168