Learn More
Medchemexpress LLC Agn 192403 hydrochloride | 1021868-90-5 | 99.0% | 189.73 g/mol | C10H20ClN | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101374A5MG
Encompass_Preferred
AGN 192403 hydrochloride is the hydrochloride salt of a selective I1-imidazoline receptor antagonist used in cardiovascular and neurological research. It is supplied in small research quantities with high reported purity, and includes solubility and formulation guidance for in vitro and in vivo studies.
  • Selective I1-imidazoline receptor antagonist for receptor pharmacology studies.
  • Hydrochloride salt form improves solubility in polar solvents.
  • High purity suitable for biochemical and cellular assays.
  • Available in small milligram quantities for preclinical research.
  • Soluble in DMSO; in vivo formulation recipes and concentrations are reported.
Catalog No. 50-002-11811