Learn More
Medchemexpress LLC AGO2/Argonaute-2 Mo 10ug
SDP
Supplier:  Medchemexpress LLC HYP74423Y10UG
Encompass_Preferred
The AGO2/Argonaute-2 protein is critical for RNA-mediated silencing complex (RISC)-mediated gene silencing In minimal RISC AGO2 binds to short guide RNAs such as microRNAs (miRNAs) or short interfering RNAs (siRNAs) to guide RISC for silencing with complementary mRNAs AGO2/Argonaute-2 Protein Mouse (sf9 His solution) is the recombinant mouse-derived AGO2/Argonaute-2 protein expressed by Sf9 insect cells with N-His labeled tag
Catalog No. 50-001-78677