Learn More
Medchemexpress LLC Agomelatine-d6 | 1079389-42-6 | MFCD21363446 | 98.6% | 249.34 g/mol | C15H11D6NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17038S10MG
Encompass_Preferred
Agomelatine-d6 is a deuterium-labeled analogue of agomelatine provided as a research-grade isotopically labeled reference standard. It is intended for use in analytical quantification, method development, and pharmacokinetic studies targeting melatonin MT1/MT2 receptor activity and 5-HT2C-related pathways. Not for human or clinical use.
  • Isotopically labeled internal standard for LC-MS/MS assays.
  • Supports accurate quantification of agomelatine in biological matrices.
  • High purity suitable for analytical and pharmacological studies.
  • Available in milligram quantities for method development and validation.
  • Supplied for research use only; not for clinical administration.
Catalog No. 50-002-95088