Learn More
TARGETMOL CHEMICALS INC AGOMELATINE HYDROCHLORID 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6052325MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Agomelatine hydrochloride (S-20098 hydrochloride) (S-20098 hydrochloride) is a specific MT1 and MT2 receptor agonist with Ki of 0.1 nM for CHO-hMT1 0.06 nM for HEK-hMT1 0.12 nM for CHO-hMT2 and 0.27 nM for HEK-hMT2 respectively [1]. Agomelatine hydrochloride is a selective antagonist of a 5-HT2C receptor with pKis of 6.4 and 6.2 for native (porcine) and cloned human 5-HT2C receptors respectively [2]. purity: 100%
Catalog No. 50-257-9611